Anti-RPL36 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128663-100 ORB128663-200
BIRBORB128663-100EA 0 EUR
BIRBORB128663-100 BIRBORB128663-200
Anti-RPL36 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RPL36 (ribosomal protein L36)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR