Anti-RRM2B Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128688-100 ORB128688-200
BIRBORB128688-100EA 0 EUR
BIRBORB128688-100 BIRBORB128688-200
Anti-RRM2B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RRM2B (ribonucleotide reductase M2 B (TP53 inducible))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR