Anti-DGKI Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128969-100 ORB128969-200
BIRBORB128969-100EA 0 EUR
BIRBORB128969-100 BIRBORB128969-200
Anti-DGKI Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DGKI (diacylglycerol kinase, iota)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR