Anti-LCP2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB135833-100 ORB135833-200 ORB135833-50
BIRBORB135833-100EA 0 EUR
BIRBORB135833-100 BIRBORB135833-200 BIRBORB135833-50
Anti-LCP2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LCP2 (lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of 76kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR