Anti-LMNB1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB135851-100 ORB135851-200 ORB135851-50
BIRBORB135851-100EA 0 EUR
BIRBORB135851-100 BIRBORB135851-200 BIRBORB135851-50
Anti-LMNB1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LMNB1 (lamin B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR