Anti-HSP90AA1 Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB67311-200 ORB67311-50
BIRBORB67311-200EA 0 EUR
BIRBORB67311-200 BIRBORB67311-50
Anti-HSP90AA1 Mouse Monoclonal Antibody
Antibodies
Western Blot: 1 ?g/ml

Type: Primary
Antigen: HSP90AA1 (heat shock protein 90kDa alpha (cytosolic), class A member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR