Anti-SRF Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB99334-100 ORB99334-20
BIRBORB99334-100EA 0 EUR
BIRBORB99334-100 BIRBORB99334-20
Anti-SRF Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: SRF (serum response factor (c-fos serum response element-binding transcription factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR