Anti-IgG Goat Polyclonal Antibody (CF™ 680)

Supplier: Biotium
20064 20064-1
20064.EA 289 EUR
20064. 20064-1
Anti-IgG Goat Polyclonal Antibody (CF™ 680)
Antibodies
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-rabbit IgG (H+L) with CF™680 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Far-red/near-infrared CF™680 (Ex/Em 681/698 nm) is brighter and provides the best signal-to-noise ratio compared to antibody conjugates of spectrally similar dyes.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF680
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR