Anti-IgG Goat Polyclonal Antibody (CF™ 640R)

Supplier: Biotium
20087 20087-1
20087.EA 206 EUR
20087. 20087-1
Anti-IgG Goat Polyclonal Antibody (CF™ 640R)
Antibodies
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-rabbit IgG (H+L) with CF™640R dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Far-red fluorescent CF™640R (Ex/Em 642/662 nm) is much brighter than Cy®5 and at least as bright as Alexa Fluor® 647, and has exceptional photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF640R
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

SPECIFICATIONS

Antigen symbol IgG
Antigen name Rabbit IgG
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation CF® 640R
Reactivity Rabbit
Host Goat
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antibody fragment F(ab')2
Form Liquid
Purification Affinity purified
Concentration 2 mg/mL
Cross adsorption No
Storage buffer PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
Storage temperature Store at –20 °C.
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR