Anti-IgG Donkey Polyclonal Antibody (CF™ 568)

Supplier: Biotium
20095 20095-1
20095.EA 359 EUR
20095. 20095-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 568)
Antibodies
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 568 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR