Anti-IgG Goat Polyclonal Antibody (CF™ 568)

Supplier: Biotium
20099 20099-1
20099.EA 238 EUR
20099. 20099-1
Anti-IgG Goat Polyclonal Antibody (CF™ 568)
Antibodies
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-rabbit IgG (H+L) with CF™568 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Red fluorescent CF™568 (Ex/Em 562/583 nm) produces much brighter antibody conjugates than Alexa Fluor® 568, and has superior photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF568
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR