Anti-CFC1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-1598 NB100-1598SS
NOVUNB100-1598EA 639 EUR
NOVUNB100-1598 NOVUNB100-1598SS
Anti-CFC1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CFC1 (cripto, FRL-1, cryptic family 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR