Anti-CHEK1 Mouse Monoclonal Antibody [clone: 2G1D5]

Supplier: Novus Biologicals
NB100-1768 NB100-1768SS
NOVUNB100-1768EA 586 EUR
NOVUNB100-1768 NOVUNB100-1768SS
Anti-CHEK1 Mouse Monoclonal Antibody [clone: 2G1D5]
Antibodies
Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: monoclonal
Clone: 2G1D5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR