Anti-TRA Mouse Monoclonal Antibody [clone: TRA-1-81]

Supplier: Novus Biologicals
NB100-1833 NB100-1833SS
NOVUNB100-1833EA 554 EUR
NOVUNB100-1833 NOVUNB100-1833SS
Anti-TRA Mouse Monoclonal Antibody [clone: TRA-1-81]
Antibodies
Type: Primary
Antigen: TRA (T cell receptor alpha locus)
Clonality: monoclonal
Clone: TRA-1-81
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR