Anti-PDI Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-1921 NB100-1921SS
NOVUNB100-1921EA 606 EUR
NOVUNB100-1921 NOVUNB100-1921SS
Anti-PDI Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDI
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR