Anti-ERO1L Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-2525 NB100-2525SS
NOVUNB100-2525EA 615 EUR
NOVUNB100-2525 NOVUNB100-2525SS
Anti-ERO1L Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ERO1L (ERO1-like (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR