Anti-4-amino Biphenyl DNA Mouse Monoclonal Antibody [clone: 4C11]

Supplier: Novus Biologicals
NB100-415 NB100-415SS
NOVUNB100-415EA 554 EUR
NOVUNB100-415 NOVUNB100-415SS
Anti-4-amino Biphenyl DNA Mouse Monoclonal Antibody [clone: 4C11]
Antibodies
Type: Primary
Antigen: 4-amino Biphenyl DNA
Clonality: monoclonal
Clone: 4C11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR