Anti-PLIN3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-40765 NB110-40765SS
NOVUNB110-40765EA 554 EUR
NOVUNB110-40765 NOVUNB110-40765SS
Anti-PLIN3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PLIN3 (perilipin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR