Anti-KIAA1524 Mouse Monoclonal Antibody [clone: 2G10]

Supplier: Novus Biologicals
NB110-59722 NB110-59722SS
NOVUNB110-59722EA 623 EUR
NOVUNB110-59722 NOVUNB110-59722SS
Anti-KIAA1524 Mouse Monoclonal Antibody [clone: 2G10]
Antibodies
Type: Primary
Antigen: KIAA1524 (KIAA1524)
Clonality: monoclonal
Clone: 2G10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR