Anti-SIAH2 Mouse Monoclonal Antibody [clone: 24E6H3]

Supplier: Novus Biologicals
NB110-88113 NB110-88113SS
NOVUNB110-88113EA 543 EUR
NOVUNB110-88113 NOVUNB110-88113SS
Anti-SIAH2 Mouse Monoclonal Antibody [clone: 24E6H3]
Antibodies
Type: Primary
Antigen: SIAH2 (siah E3 ubiquitin protein ligase 2)
Clonality: monoclonal
Clone: 24E6H3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR