Anti-S100A12 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-90136 NB110-90136SS
NOVUNB110-90136EA 554 EUR
NOVUNB110-90136 NOVUNB110-90136SS
Anti-S100A12 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: S100A12 (S100 calcium binding protein A12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR