Anti-SERPINB5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-97377 NB110-97377SS
NOVUNB110-97377EA 554 EUR
NOVUNB110-97377 NOVUNB110-97377SS
Anti-SERPINB5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SERPINB5 (serpin peptidase inhibitor, clade B (ovalbumin), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR