Anti-FXR2 Mouse Monoclonal Antibody [clone: 1G2]

Supplier: Novus Biologicals
NB300-259 NB300-259SS
NOVUNB300-259EA 554 EUR
NOVUNB300-259 NOVUNB300-259SS
Anti-FXR2 Mouse Monoclonal Antibody [clone: 1G2]
Antibodies
Type: Primary
Antigen: FXR2 (fragile X mental retardation, autosomal homolog 2)
Clonality: monoclonal
Clone: 1G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR