Anti-Endothelial Lipase Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB400-118 NB400-118SS
NOVUNB400-118EA 612 EUR
NOVUNB400-118 NOVUNB400-118SS
Anti-Endothelial Lipase Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Endothelial Lipase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR