Anti-APOA5 Mouse Monoclonal Antibody [clone: 1G5G9]

Supplier: Novus Biologicals
NB400-139 NB400-139SS
NOVUNB400-139EA 536 EUR
NOVUNB400-139 NOVUNB400-139SS
Anti-APOA5 Mouse Monoclonal Antibody [clone: 1G5G9]
Antibodies
Type: Primary
Antigen: APOA5 (apolipoprotein A-V)
Clonality: monoclonal
Clone: 1G5G9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR