Anti-CD4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-19371 NBP1-19371SS
NOVUNBP1-19371EA 732 EUR
NOVUNBP1-19371 NOVUNBP1-19371SS
Anti-CD4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR