Anti-NANOG Mouse Monoclonal Antibody [clone: 1E6C4]

Supplier: Novus Biologicals
NBP1-47427 NBP1-47427SS
NOVUNBP1-47427EA 566 EUR
NOVUNBP1-47427 NOVUNBP1-47427SS
Anti-NANOG Mouse Monoclonal Antibody [clone: 1E6C4]
Antibodies
Type: Primary
Antigen: NANOG (Nanog homeobox)
Clonality: monoclonal
Clone: 1E6C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR