Anti-KDM4A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-49602 NBP1-49602SS
NOVUNBP1-49602EA 554 EUR
NOVUNBP1-49602 NOVUNBP1-49602SS
Anti-KDM4A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal JMJD2A Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50

Type: Primary
Antigen: JMJD2A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR