Anti-GPR155 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-50625 NBP1-50625SS
NOVUNBP1-50625EA 554 EUR
NOVUNBP1-50625 NOVUNBP1-50625SS
Anti-GPR155 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GPR155 (G protein-coupled receptor 155)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR