Anti-HTR4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-78403 NBP1-78403SS
NOVUNBP1-78403EA 446 EUR
NOVUNBP1-78403 NOVUNBP1-78403SS
Anti-HTR4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HTR4 (5-hydroxytryptamine (serotonin) receptor 4, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR