Anti-IDH2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-22166 NBP2-22166SS
NOVUNBP2-22166EA 554 EUR
NOVUNBP2-22166 NOVUNBP2-22166SS
Anti-IDH2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IDH2 (isocitrate dehydrogenase 2 (NADP+), mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR