Anti-GLI2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-23602 NBP2-23602SS
NOVUNBP2-23602EA 606 EUR
NOVUNBP2-23602 NOVUNBP2-23602SS
Anti-GLI2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GLI2 (GLI family zinc finger 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR