Anti-FOXE3 Rabbit Polyclonal Antibody

Supplier: US Biological
F9046-31-100 F9046-31-50
USBIF9046-31-100EA 270 EUR
USBIF9046-31-100 USBIF9046-31-50
Anti-FOXE3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1: 250000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: FOXE3
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR