Anti-IGFBP1 Rabbit Polyclonal Antibody

Supplier: US Biological
143591 143591-100
USBI143591EA 473 EUR
USBI143591 USBI143591-100
Anti-IGFBP1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 0.5-2?g/ml

Type: Primary
Antigen: IGFBP1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR