Anti-MART-1 Mouse Monoclonal Antibody [clone: M2-7C10]

Supplier: US Biological
138568 138568-100
USBI138568EA 391 EUR
USBI138568 USBI138568-100
Anti-MART-1 Mouse Monoclonal Antibody [clone: M2-7C10]
Antibodies
Recommended dilution: ;Western Blot: 0.5-1 ?g/ml; Immunofluorescence: 1-2?g/ml; Immunoprecipitation: 1-2?g/500 ?g; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: MART-1
Clonality: Monoclonal
Clone: M2-7C10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR