Polyvinyl alcohol, M.W. ∼ 160000 for synthesis, partially hydrolysed, Sigma-Aldrich®


M-Clarity™ quality level = MQ200

8.43870.0100 8.43870.1000
8.43870.0100EA 50.2 EUR
8.43870.0100 8.43870.1000
Polyvinyl alcohol, M.W. ∼ 160000 for synthesis, partially hydrolysed, Sigma-Aldrich®
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Solubility Insoluble at 20 °C
Melting Point 200 °C
Bulk Density 400 - 600 kg/m3
Density 1.3 g/cm3 (20 °C)
pH 4.5 - 7.0 (40 g/l, H2O, 23 °C)
Ignition Point 230 °C
Bulk density 400 - 600 kg/m3
Storage temperature 15...25 °C
Degree of hydrolysis ≥85%
dyn.Viscosity 22,1 - 29,9 mPa∙s
Loss on drying (110 °C, 2 h) ≤5,0%
Ash ≤1,0%
Ester value 130 - 150

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR