Bromopentacarbonylmanganese(I) ≥98%
|
Danger
|
39310.03
39310.06
39310.03EA
100
EUR
39310.03
39310.06
Bromopentacarbonylmanganese(I) ≥98%
Bromopentacarbonylmanganese(I)
Purity: 98%
CAS No.: 14516-54-2
Molecular Formula: C5BrMnO5
Formula Weight: 274.89
Physical Form: Crystalline
Sensitivity: Air and Moisture Sensitive
Appearance: Orange
UN No.: UN3466
MDL No.: MFCD00049806
Formula:
C₅BrMnO₅ MW: 274.89 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00049806 CAS Number: 14516-54-2 EINECS: 238-522-8 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...