Silver tungsten oxide ≥99%

Supplier: ALFA AESAR

39661.09 39661.18
39661.09EA 97.2 EUR
39661.09 39661.18
Silver tungsten oxide ≥99%
Silver tungsten oxide

Purity: 99% CAS No.: 13465-93-5 Molecular Formula: Ag2O4W Formula Weight: 463.58 Physical Form: Powder Sensitivity: Light Sensitive Appearance: Light yellow Odour: Odourless Solubility: Insoluble in water. Soluble in potassium cyanide, ammonium hydroxide and nitric acid MDL No.: MFCD00014147

Formula: Ag₂O₄W
Storage Temperature: Ambient
MDL Number: MFCD00014147
CAS Number: 13465-93-5
EINECS: 236-708-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR