Ethylenediaminetetraacetic acid tetrakis(tetramethylammonium) salt 30% (w/v) in aqueous solution
Supplier: ALFA AESAR
41518.K2
41518.AD
41518.K2EA
0
EUR
41518.K2
41518.AD
Ethylenediaminetetraacetic acid tetrakis(tetramethylammonium) salt 30% (w/v) in aqueous solution
Ethylenediaminetetraacetic acid tetrakis(tetramethylammonium) salt
Molecular Formula: C26H60N6O8
Formula Weight: 276.44
Physical Form: Liquid
MDL No.: MFCD01632148
Applications: Chelating agent, polymerization catalyst, detergent
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...