Platinum 10% (nominal) on carbon black, HiSPEC® 2000
|
Danger
|
43876.02
43876.09
43876.02EA
81
EUR
43876.02
43876.09
Platinum 10% (nominal) on carbon black, HiSPEC® 2000
Platinum
CAS No.: 7440-06-4
Odour: Odourless
UN No.: UN1325
MDL No.: MFCD00011179
Formula:
Pt MW: 195.08 g/mol Boiling Pt: 3827 Melting Pt: 1769 Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 UN: 1325 ADR: 4.1 II Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...