2-Hydroxypyrimidine hydrochloride ≥98%
A13732.14
A13732.22
A13732.36
A13732.14EA
56.6
EUR
A13732.14
A13732.22
A13732.36
2-Hydroxypyrimidine hydrochloride ≥98%
2-Hydroxypyrimidine hydrochloride
Purity: 98+%
CAS No.: 38353-09-2
Molecular Formula: C4H4N2O.HCl
Formula Weight: 132.55
Melting Point: ca 210° dec.
Beilstein Reference: 3560963
MDL No.: MFCD00012781
Formula:
C₄H₄N₂O Melting Pt: ∼ 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012781 CAS Number: 38353-09-2 EINECS: 253-897-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...