Pivaloylacetonitrile 99%

Supplier: ALFA AESAR
Danger

A16309.14 A16309.22
A16309.14EA 72.4 EUR
A16309.14 A16309.22
Pivaloylacetonitrile 99%
Pivaloylacetonitrile

Purity: 99% CAS No.: 59997-51-2 Molecular Formula: C7H11NO Formula Weight: 125.17 Melting Point: 66-70° Beilstein Reference: 1746674 UN No.: UN3439 MDL No.: MFCD00010208

Formula: C₇H₁₁NO
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00010208
CAS Number: 59997-51-2
EINECS: 262-017-1
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR