Aluminium oxide activated neutral γ-phase ≥99.9% (metals basis)

43266.22 43266.36
43266.22EA 94.5 EUR
43266.22 43266.36
Aluminium oxide activated neutral γ-phase ≥99.9% (metals basis)
Aluminium oxide activated neutral

Purity: 99.9% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 40 Micron APS Powder, S.A. 100m2/g Appearance: White Odour: Odourless Merck Reference: 14,356 MDL No.: MFCD00003424

Formula: Al₂O₃
MW: 101.96 g/mol
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR