1-(6-Methyl-2-pyridyl)homopiperazine ≥99%
Supplier: ALFA AESAR
H50876.03
H50876.MD
H50876.03EA
0
EUR
H50876.03
H50876.MD
1-(6-Methyl-2-pyridyl)homopiperazine ≥99%
1-(6-Methyl-2-pyridyl)homopiperazine
Purity: 99%
CAS No.: 880361-95-5
Molecular Formula: C11H17N3
Formula Weight: 191.28
UN No.: UN3259
MDL No.: MFCD08061041
Formula:
C₁₁H₁₇N₃ |
MDL Number:
MFCD08061041 CAS Number: 880361-95-5 ADR: , |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...