Benzylpyridin-3-ylmethylamine 97%
Supplier: ALFA AESAR
|
Warning
|
H50978.03
H50978.MD
H50978.03EA
262
EUR
H50978.03
H50978.MD
Benzylpyridin-3-ylmethylamine 97%
Benzylpyridin-3-ylmethylamine
Purity: 97%
CAS No.: 63361-56-8
Molecular Formula: C13H14N2
Formula Weight: 198.27
MDL No.: MFCD08061098
Formula:
C₁₃H₁₄N₂ Storage Temperature: Ambient |
MDL Number:
MFCD00716866 CAS Number: 63361-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...