4-(1,3,3-Trimethyl-7-oxabicyclo[4.1.0]hept-2-yl)-3-buten-2-one, tech.

H53499.03 H53499.06
H53499.03EA 78 EUR
H53499.03 H53499.06
4-(1,3,3-Trimethyl-7-oxabicyclo[4.1.0]hept-2-yl)-3-buten-2-one, tech.
4-(1,3,3-Trimethyl-7-oxabicyclo[4.1.0]hept-2-yl)-3-buten-2-one

CAS No.: 190059-33-7 Molecular Formula: C13H20O2 Formula Weight: 208.30 Refractive Index: 1.491 MDL No.: MFCD07784337

Formula: C₁₃H₂₀O₂
Density: 1,039
Storage Temperature: Ambient
MDL Number: MFCD07784337
CAS Number: 190059-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR