6-Nitro-1H-indole-2-carboxylic acid 97%
Supplier: ALFA AESAR
|
Warning
|
H63032.MD
H63032.03
H63032.06
H63032.MDEA
0
EUR
H63032.MD
H63032.03
H63032.06
6-Nitro-1H-indole-2-carboxylic acid 97%
6-Nitro-1H-indole-2-carboxylic acid
Purity: 97%
CAS No.: 10242-00-9
Molecular Formula: C9H6N2O4
Formula Weight: 206.15
Physical Form: Solid
MDL No.: MFCD02664466
Formula:
C₉H₆N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD02664466 CAS Number: 10242-00-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...