Methyl-5-bromosalicylate 98+%
Supplier: ALFA AESAR
|
Warning
|
H64372.06
H64372.14
H64372.06EA
0
EUR
H64372.06
H64372.14
Methyl-5-bromosalicylate 98+%
Methyl 5-bromosalicylate
Purity: 98+%
CAS No.: 4068-76-2
Molecular Formula: C8H7BrO3
Formula Weight: 231.05
Physical Form: Solid
Appearance: White to pale brown
Melting Point: 61-65° lit.
Beilstein Reference: 3047854
MDL No.: MFCD00016460
Formula:
C₈H₇BrO₃ Melting Pt: 61…65 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00016460 CAS Number: 4068-76-2 EINECS: 223-776-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...