Methoxyacetic anhydride 95%
Supplier: ALFA AESAR
|
Danger
|
H64541.14
H64541.22
H64541.14EA
146
EUR
H64541.14
H64541.22
Methoxyacetic anhydride 95%
Methoxyacetic anhydride
Purity: 95%
CAS No.: 19500-95-9
Molecular Formula: C6H10O5
Formula Weight: 162.14
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Colourless
Beilstein Reference: 1771552
UN No.: UN3265
MDL No.: MFCD11976032
Formula:
C₆H₁₀O₅ Storage Temperature: Ambient |
MDL Number:
MFCD11976032 CAS Number: 19500-95-9 EINECS: 243-114-8 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...