Fluorolink® E10-H, Fluorolink®

Supplier: ALFA AESAR

H66374.14 H66374.22
H66374.14EA 0 EUR
H66374.14 H66374.22
Fluorolink® E10-H, Fluorolink®
Fluorolink® E10-H

Applications: Use in polymer modifications.

Density: 1,73
Storage Temperature: Ambient
CAS Number: 162492-15-1
EINECS: 500-749-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR